Assay Details
| Gene Symbol: Gprin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048836.1 | - | 1 | 2444 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101930 | AK048836.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101930 | BC066049.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121677 | NM_183209.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186701 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39650 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110554 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121677 | NM_183209.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110554 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11672095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11557398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11250415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11250410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11250419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11318966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11557388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11318974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11557373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11672094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11557387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11250399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11318960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11318971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11557400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11557374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11672093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11557345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29528540 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10418904 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418902 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443997_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10418903 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10418901 | - | - |
| Mouse Genome 430 2.0 Array | 1443997_at | - | - |
| Mouse Genome 430 2.0 Array | 1443997_PM_at | - | - |
| Mouse Expression Set 430 | 1443997_at | - | - |
| Mouse Expression Set 430 | 1443997_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443997_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443997_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 774410 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246116 | - | - |

Gprin2
Genomic Map