Assay Details
| Gene Symbol: Bcl2a1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007535.2 | NP_031561.2 | 1 | 62 | 161 | |
| GenBank mRNA | BC120720.1 | - | 1 | 62 | 40129643 | 161 |
| BC132545.1 | - | 1 | 62 | 161 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299771 | BC132545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299771 | NM_007535.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299771 | BC120720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299771 | BC132545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299771 | NM_007535.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299771 | BC120720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299771 | BC132545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299771 | NM_007535.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299771 | BC120720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299771 | BC132545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299771 | NM_007535.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299771 | BC120720.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844091 | - | - |
| Whole Mouse Genome G4122A | A_52_P299771 | BC132545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299771 | NM_007535.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299771 | BC120720.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299771 | BC132545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299771 | NM_007535.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299771 | BC120720.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844064 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10589885 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10587692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10595635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10587684 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10589884 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10595633 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10587683 | - | - |
| Mouse Genome 430 2.0 Array | 1450812_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450812_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900012669.mm.1 | NM_007535.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012669.mm.1 | NM_007535.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900012669.mm.1 | TC0900001511.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450812_at | - | - |
| Mouse Expression Set 430 | 1450812_PM_at | - | - |
| Murine Genome U74v2 Set | 93869_PM_s_at | - | - |
| Murine Genome U74v2 Set | 93869_s_at | - | - |
| Murine Genome U74v2 Set | 102914_PM_s_at | - | - |
| Murine Genome U74v2 Set | 102914_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450812_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450812_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 548782 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1400156 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1400156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400156 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2678894 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2678894 | - | - |
| mouse-6 v1.1 (Illumina) | scl0012046.1_196-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400156 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012046.1_196-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400156 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2936165 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2678894 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2936165 | - | - |

Bcl2a1c
Genomic Map