Assay Details
| Gene Symbol: Tdg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011561.2 | NP_035691.2 | 4 - 5 | 586 | 70 | |
| NM_172552.3 | NP_766140.2 | 4 - 5 | 685 | 70 | ||
| GenBank mRNA | AF102875.1 | - | 2 - 3 | 258 | 70 | |
| AK053870.1 | - | 4 - 5 | 613 | 70 | ||
| AK164090.1 | - | 4 - 5 | 546 | 70 | ||
| AK164643.1 | - | 4 - 5 | 653 | 70 | ||
| AK169768.1 | - | 4 - 5 | 609 | 70 | ||
| BC010315.1 | - | 4 - 5 | 575 | 3595866 | 70 | |
| BC085470.1 | - | 4 - 5 | 632 | 6416434 | 70 | |
| BC085471.1 | - | 4 - 5 | 656 | 6816716 | 70 | |
| Gene Symbol: Gm9855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037190.1 | 1 | 582 | 70 | ||
| GenBank mRNA | AF069519.1 | - | 1 | 543 | 70 | |
| AK089915.1 | - | 1 | 586 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tdg
Genomic Map