Assay Details
| Gene Symbol: Apbb1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019456.2 | NP_062329.2 | 12 - 13 | 1572 | 90 | |
| GenBank mRNA | AF020313.1 | - | 9 - 10 | 1177 | 90 | |
| AK089987.1 | - | 4 - 5 | 446 | 90 | ||
| BC023110.1 | - | 12 - 13 | 1461 | 3707478 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apbb1ip
Genomic Map