Assay Details
| Gene Symbol: Rpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024212.4 | NP_077174.1 | 4 - 5 | 463 | 129 | |
| GenBank mRNA | AK007625.1 | - | 4 - 5 | 477 | 129 | |
| AK008098.1 | - | 4 - 5 | 476 | 129 | ||
| AK012432.1 | - | 4 - 5 | 476 | 129 | ||
| AK050639.1 | - | 4 - 5 | 477 | 129 | ||
| AK088279.1 | - | 4 - 5 | 475 | 129 | ||
| AK151950.1 | - | 4 - 5 | 473 | 129 | ||
| AK165962.1 | - | 4 - 5 | 477 | 129 | ||
| AK166748.1 | - | 4 - 5 | 476 | 129 | ||
| AK166957.1 | - | 4 - 5 | 480 | 129 | ||
| AK167054.1 | - | 4 - 5 | 478 | 129 | ||
| BC003459.1 | - | 4 - 5 | 460 | 2648518 | 129 | |
| BC094036.1 | - | 4 - 5 | 458 | 5011554 | 129 | |
| BC132321.1 | - | 4 - 5 | 463 | 40130598 | 129 | |
| BC132323.1 | - | 4 - 5 | 463 | 40130600 | 129 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107386 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141968 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014807 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P376829 | BC132323.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P376829 | AK167054.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P376829 | BC003459.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P376829 | BC094036.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P376829 | BC132321.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580549 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06818584 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141968 | - | - |
| Whole Mouse Genome G4122A | A_52_P376829 | BC132323.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P376829 | AK167054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P376829 | BC003459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P376829 | BC094036.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P376829 | BC132321.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129242 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07653144 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07653118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08086660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08086663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07653108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26580532 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10586160 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586164 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586161 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586159 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586163 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586158 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586162 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425183_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10586157 | - | - |
| Mouse Genome 430 2.0 Array | 1425183_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425183_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67891_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67891_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900006074.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006074.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006074.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006072.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006072.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006072.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006079.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006079.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006079.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006070.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006070.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006070.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006080.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006080.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006080.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006077.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006077.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006077.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006073.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006073.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006073.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006076.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006076.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006076.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006075.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006075.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006075.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006078.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006078.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006078.mm.1 | TC0900000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006071.mm.1 | NM_024212.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006071.mm.1 | NM_024212.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006071.mm.1 | TC0900000810.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425183_a_at | - | - |
| Mouse Expression Set 430 | 1425183_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425183_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425183_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425183_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425183_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 353265 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5420324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5420324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2940014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420324 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2725863 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631003 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2725863 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2631003 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067891.1_266-S | - | - |
| mouse-6 v1.1 (Illumina) | scl36833.9.1_209-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940014 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420324 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36833.9.1_209-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067891.1_266-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420324 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2631003 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2874507 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631003 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2874507 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36833.9.1_209-S | - | - |

Rpl4
Genomic Map