Assay Details
| Gene Symbol: 3300002I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027017.1 | NP_081293.4 | 2 - 3 | 303 | 234 | |
| GenBank mRNA | AK014369.1 | - | 2 - 3 | 303 | 234 | |
| BC132262.1 | - | 2 - 3 | 244 | 40130539 | 234 | |
| BC132264.1 | - | 2 - 3 | 244 | 40130541 | 234 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006479 | NM_027017.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006479 | AK014369.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473372 | AK014103.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473372 | AK077954.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473372 | AK081763.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473372 | AK148294.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473372 | XM_001001171.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473372 | XM_912718.3 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394830 | AK044557.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167024 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119342 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006479 | NM_027017.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006479 | AK014369.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21739137 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04737272 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04737317 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04737282 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04737290 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04737304 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00525972 | - | - |
| Whole Mouse Genome G4122A | A_51_P147213 | NM_027017.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147213 | AK013537.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147213 | AK014369.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04737282 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04737304 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02186197 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02186195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02186185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02186186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21739004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21738910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01422463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02148620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01290198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21739137 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488453 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432534_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488451 | - | - |
| Mouse Genome 430 2.0 Array | 1432534_at | - | - |
| Mouse Genome 430 2.0 Array | 1445154_at | - | - |
| Mouse Genome 430 2.0 Array | 1445154_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432534_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69277_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69277_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200041169.mm.1 | NM_027017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041169.mm.1 | NM_027017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041169.mm.1 | TC0200004833.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041170.mm.1 | NM_027017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041170.mm.1 | NM_027017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041170.mm.1 | TC0200004833.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041174.mm.1 | NM_027017.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041174.mm.1 | NM_027017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041174.mm.1 | TC0200004833.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432534_at | - | - |
| Mouse Expression Set 430 | 1432534_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432534_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432534_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 489561 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4560338 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4560338 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560338 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238502 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238502 | - | - |
| mouse-6 v1.1 (Illumina) | scl18531.6.1_97-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560338 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18531.6.1_97-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560338 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1238502 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238502 | - | - |

3300002I08Rik
Genomic Map