| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100001617.mm.1 |
NM_033523.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100001617.mm.1 |
NM_033523.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100001617.mm.1 |
TC1100000187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100001622.mm.1 |
NM_033523.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100001622.mm.1 |
NM_033523.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100001622.mm.1 |
TC1100000187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100001616.mm.1 |
TC1100000187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100001619.mm.1 |
NM_033523.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100001619.mm.1 |
NM_033523.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100001619.mm.1 |
TC1100000187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100001621.mm.1 |
NM_033523.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100001621.mm.1 |
NM_033523.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100001621.mm.1 |
TC1100000187.mm.1 |
MTA TC |