Assay Details
| Gene Symbol: Vmn1r216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134245.1 | NP_599006.1 | 1 | 360 | 189 | |
| GenBank mRNA | BC145650.1 | - | 1 | 438 | 40130953 | 189 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203306 | NM_134245.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203306 | BC145650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203306 | NM_134245.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203306 | BC145650.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132613 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203306 | NM_134245.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203306 | BC145650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203306 | NM_134245.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203306 | BC145650.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203306 | NM_134245.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203306 | BC145650.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203306 | NM_134245.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203306 | BC145650.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879890 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132613 | - | - |
| Whole Mouse Genome G4122A | A_51_P203306 | NM_134245.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203306 | BC145650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203306 | NM_134245.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203306 | BC145650.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140265 | NM_001083311.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128886 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132613 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10901597 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10901592 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10901602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10901604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10581192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10667401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10982717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10985532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10536011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10674434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10982721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10581196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10982715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10778801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10735580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879915 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404023 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404022 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404014 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300001326.mm.1 | TC1300000226.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001328.mm.1 | TC1300000226.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001327.mm.1 | NM_134245.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001327.mm.1 | NM_134245.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001327.mm.1 | TC1300000226.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 511196 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4230086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4230086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004230086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2419707 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2419707 | - | - |
| mouse-6 v1.1 (Illumina) | scl12339.1.1_40-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4230086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl12339.1.1_40-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4230086 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2934639 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2419707 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2934639 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2419707 | - | - |

Vmn1r216
Genomic Map