Assay Details
| Gene Symbol: Olfr1484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146291.1 | NP_666403.1 | 1 | 344 | 186 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252586 | NM_146291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252586 | NM_146291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252586 | NM_146291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252586 | NM_146291.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P252586 | NM_146291.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P252586 | NM_146291.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965091 | - | - |
| Whole Mouse Genome G4122A | A_51_P252586 | NM_146291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252586 | NM_146291.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14271630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14271635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14497038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14497041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965115 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461817 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461816 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900003042.mm.1 | NM_146291.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003042.mm.1 | NM_146291.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003042.mm.1 | TC1900000277.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 696802 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3780164 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3780164 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780164 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2663722 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2663722 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258288.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780164 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258288.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780164 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2663722 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2663722 | - | - |

Olfr1484
Genomic Map