Assay Details
| Gene Symbol: Olfr584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147054.1 | NP_667265.1 | 1 | 438 | 68 | |
| GenBank mRNA | BC128007.1 | - | 1 | 442 | 40135987 | 68 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P309804 | BC128007.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P309804 | NM_147054.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P309804 | BC128007.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P309804 | NM_147054.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P309804 | BC128007.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P309804 | NM_147054.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P309804 | BC128007.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P309804 | NM_147054.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P309805 | BC128007.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P309805 | NM_147054.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P309804 | BC128007.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P309804 | NM_147054.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P309804 | BC128007.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P309804 | NM_147054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309804 | BC128007.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309804 | NM_147054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309804 | BC128007.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309804 | NM_147054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309805 | BC128007.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P309805 | NM_147054.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06200800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06161424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06428824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06177355 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555763 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555762 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700012716.mm.1 | NM_147054.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012716.mm.1 | NM_147054.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012716.mm.1 | TC0700001476.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 469029 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6590102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590102 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739006 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2739006 | - | - |
| mouse-6 v1.1 (Illumina) | scl0259056.1_328-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0259056.1_328-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590102 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2739006 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739006 | - | - |

Olfr584
Genomic Map