Assay Details
| Gene Symbol: Epn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252188.1 | NP_001239117.1 | 6 - 7 | 1352 | 102 | |
| NM_001252189.1 | NP_001239118.1 | 4 - 5 | 1108 | 102 | ||
| NM_010148.3 | NP_034278.1 | 5 - 6 | 1144 | 102 | ||
| GenBank mRNA | AF057286.1 | - | 3 - 4 | 599 | 102 | |
| AK036331.1 | - | 4 - 5 | 1082 | 102 | ||
| AK139234.1 | - | 6 - 7 | 1340 | 102 | ||
| AK139793.1 | - | 6 - 7 | 1275 | 102 | ||
| AK160782.1 | - | 4 - 5 | 1109 | 102 | ||
| AK173093.1 | - | 6 - 7 | 1328 | 102 | ||
| BC039138.1 | - | 5 - 6 | 1108 | 2647379 | 102 | |
More Information
Set Membership: |
|

Epn2
Genomic Map