Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 4 - 5 | 442 | 59 | |
| GenBank mRNA | AK140597.1 | - | 4 - 5 | 463 | 59 | |
| AK150939.1 | - | 4 - 5 | 462 | 59 | ||
| AK156079.1 | - | 4 - 5 | 412 | 59 | ||
| AK171348.1 | - | 4 - 5 | 458 | 59 | ||
| AK183517.1 | - | 2 - 3 | 186 | 59 | ||
| AK183726.1 | - | 2 - 3 | 147 | 59 | ||
| AK184446.1 | - | 2 - 3 | 186 | 59 | ||
| AK196904.1 | - | 2 - 3 | 186 | 59 | ||
| AK210357.1 | - | 2 - 3 | 182 | 59 | ||
| BC015255.1 | - | 4 - 5 | 442 | 4221225 | 59 | |
| BC139818.1 | - | 4 - 5 | 390 | 40141268 | 59 | |
| CT010198.1 | - | 4 - 5 | 246 | 59 | ||
| D87911.1 | - | 4 - 5 | 365 | 59 | ||
| U60330.1 | - | 4 - 5 | 246 | 59 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map