Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 6 - 7 | 598 | 61 | |
| GenBank mRNA | AK140597.1 | - | 6 - 7 | 619 | 61 | |
| AK150939.1 | - | 6 - 7 | 618 | 61 | ||
| AK156079.1 | - | 6 - 7 | 568 | 61 | ||
| AK171348.1 | - | 6 - 7 | 614 | 61 | ||
| AK178475.1 | - | 5 - 6 | 319 | 61 | ||
| AK183517.1 | - | 4 - 5 | 342 | 61 | ||
| AK183726.1 | - | 4 - 5 | 306 | 61 | ||
| AK210357.1 | - | 4 - 5 | 338 | 61 | ||
| BC015255.1 | - | 6 - 7 | 598 | 4221225 | 61 | |
| BC139818.1 | - | 6 - 7 | 546 | 40141268 | 61 | |
| CT010198.1 | - | 6 - 7 | 402 | 61 | ||
| D87911.1 | - | 6 - 7 | 521 | 61 | ||
| U60330.1 | - | 6 - 7 | 402 | 61 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map