Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 7 - 8 | 667 | 93 | |
| GenBank mRNA | AK140597.1 | - | 7 - 8 | 688 | 93 | |
| AK150939.1 | - | 7 - 8 | 687 | 93 | ||
| AK156079.1 | - | 7 - 8 | 637 | 93 | ||
| AK171348.1 | - | 7 - 8 | 683 | 93 | ||
| AK178475.1 | - | 6 - 7 | 388 | 93 | ||
| BC015255.1 | - | 7 - 8 | 667 | 4221225 | 93 | |
| BC139818.1 | - | 7 - 8 | 615 | 40141268 | 93 | |
| CT010198.1 | - | 7 - 8 | 471 | 93 | ||
| D87911.1 | - | 7 - 8 | 590 | 93 | ||
| U60330.1 | - | 7 - 8 | 471 | 93 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map