Assay Details
| Gene Symbol: Serpina1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009246.3 | NP_033272.1 | 3 - 4 | 1005 | 105 | |
| GenBank mRNA | AK002537.1 | - | 3 - 4 | 965 | 105 | |
| M75718.1 | - | 3 - 4 | 946 | 105 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of peptidase activity
negative regulation of endopeptidase activity
response to cytokine
response to peptide hormone
glycoprotein binding
protease binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity
identical protein binding
negative regulation of peptidase activity
negative regulation of endopeptidase activity
response to cytokine
response to peptide hormone
glycoprotein binding
protease binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010298 | M75718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010298 | AK002537.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010298 | NM_009246.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010298 | BC021850.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121117 | M75718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121117 | AK002537.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121117 | NM_009246.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121117 | BC021850.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010298 | M75718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010298 | AK002537.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010298 | NM_009246.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010298 | BC021850.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121117 | M75718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121117 | AK002537.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121117 | NM_009246.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121117 | BC021850.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236303 | M75718.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236303 | NM_009246.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236303 | M75718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236303 | NM_009246.3 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28699485 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10402403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402401 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402410 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402411 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402402 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402400 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402413 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10402399 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10402394 | - | - |
| Mouse Genome 430 2.0 Array | 1449321_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1449321_PM_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200015832.mm.1 | NM_009246.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015832.mm.1 | NM_009246.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015832.mm.1 | TC1200002532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200015835.mm.1 | NM_009246.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015835.mm.1 | NM_009246.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015835.mm.1 | TC1200002532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200015833.mm.1 | NM_009246.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015833.mm.1 | NM_009246.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015833.mm.1 | TC1200002532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200015831.mm.1 | NM_009246.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015831.mm.1 | NM_009246.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015831.mm.1 | TC1200002532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200015834.mm.1 | NM_009246.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015834.mm.1 | NM_009246.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015834.mm.1 | TC1200002532.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449321_x_at | - | - |
| Mouse Expression Set 430 | 1449321_PM_x_at | - | - |
| Murine Genome U74v2 Set | 93109_f_at | - | - |
| Murine Genome U74v2 Set | 101565_f_at | - | - |
| Murine Genome U74v2 Set | 93109_PM_f_at | - | - |
| Murine Genome U74v2 Set | 101565_PM_f_at | - | - |
| Murine Genome U74v2 Set | 101575_i_at | - | - |
| Murine Genome U74v2 Set | 101576_f_at | - | - |
| Murine Genome U74v2 Set | 101575_PM_i_at | - | - |
| Murine Genome U74v2 Set | 101576_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449321_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449321_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3120692 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3120692 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003120692 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225570 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225570 | - | - |
| mouse-6 v1.1 (Illumina) | scl020703.2_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3120692 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020703.2_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3120692 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225570 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225570 | - | - |

Serpina1d
Genomic Map