Assay Details
| Gene Symbol: B930041F14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178699.4 | NP_848814.3 | 1 | 2083 | 154 | |
| GenBank mRNA | AB041660.1 | - | 1 | 1604 | 154 | |
| AK046537.1 | - | 1 | 2086 | 154 | ||
| AK047240.1 | - | 1 | 1954 | 154 | ||
| AK048841.1 | - | 1 | 1928 | 154 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440035 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440037 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440043 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170732 | NM_178699.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170732 | AB041660.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012734 | AK046537.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012734 | AK047240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012734 | NM_178699.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012734 | AB041660.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160643 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105451 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012734 | AK046537.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012734 | AK047240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012734 | NM_178699.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012734 | AB041660.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170732 | NM_178699.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170732 | AB041660.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440050 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105451 | - | - |
| Whole Mouse Genome G4122A | A_51_P381086 | AK046537.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381086 | AK047240.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381086 | AK048841.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381086 | NM_178699.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381086 | AB041660.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132665 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105451 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03904587 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03945167 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03904588 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440037 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440035 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03904591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03904588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03906276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03948350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03945167 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10511138 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438635_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427230_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10511136 | - | - |
| Mouse Genome 430 2.0 Array | 1438635_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438635_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427230_at | - | - |
| Mouse Genome 430 2.0 Array | 1427230_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230991_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230991_at | - | - |
| Mouse Expression Set 430 | 1427230_PM_at | - | - |
| Mouse Expression Set 430 | 1427230_at | - | - |
| Mouse Expression Set 430 | 1438635_PM_x_at | - | - |
| Mouse Expression Set 430 | 1438635_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427230_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427230_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427230_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427230_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438635_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438635_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 740356 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1660348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250201 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250201 | - | - |
| mouse-6 v1.1 (Illumina) | scl24616.1.1396_49-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24616.1.1396_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1250201 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250201 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24616.1.1396_49-S | - | - |

B930041F14Rik
Genomic Map