Assay Details
Gene Symbol: Olfr1250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146965.1 | NP_667176.1 | 1 | 548 | 148 | |
GenBank mRNA | BC125323.1 | - | 1 | 642 | 40129838 | 148 |
BC125349.1 | - | 1 | 642 | 40129864 | 148 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028842 | NM_146965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028842 | BC125323.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028842 | BC125349.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028842 | NM_146965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028842 | BC125323.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028842 | BC125349.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160143 | NM_146965.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160143 | BC125323.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160143 | BC125349.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160143 | NM_146965.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160143 | BC125323.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160143 | BC125349.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414466 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02259435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414465 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01869435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414518 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414469 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414484 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414490 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414461 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414516 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484842 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484843 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484841 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034090.mm.1 | TC0200004068.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200034091.mm.1 | NM_146965.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034091.mm.1 | NM_146965.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034091.mm.1 | TC0200004068.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200034092.mm.1 | TC0200004068.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 681217 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102850398 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1400010 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001400010 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255403 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1255403 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1250968 | - | - |
mouse-6 v1.1 (Illumina) | GI_22128996-S | - | - |
mouse-6 v1.1 (Illumina) | scl0258967.1_69-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1400010 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102850398 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258967.1_69-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102850398 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400010 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1255403 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255403 | - | - |
