Assay Details
| Gene Symbol: Fhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077361.1 | NP_001070829.1 | 6 - 7 | 985 | 61 | |
| GenBank mRNA | AK158966.1 | - | 6 - 7 | 913 | 61 | |
| BC055725.1 | - | 1 - 2 | 57 | 61 | ||
| BC059009.1 | - | 3 - 4 | 1265 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fhl1
Genomic Map