Assay Details
| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011074.2 | NP_035204.2 | 11 - 12 | 1230 | 91 | |
| GenBank mRNA | AF033655.1 | - | 11 - 12 | 1189 | 91 | |
| AK129226.1 | - | 11 - 12 | 1057 | 91 | ||
| AK138437.1 | - | 10 - 11 | 1471 | 91 | ||
| AK139386.1 | - | 10 - 11 | 1391 | 91 | ||
| AK141535.1 | - | 10 - 11 | 1159 | 91 | ||
| AK165149.1 | - | 10 - 11 | 1075 | 91 | ||
| BC068134.1 | - | 10 - 11 | 1434 | 6834018 | 91 | |
| U62391.1 | - | 10 - 11 | 1084 | 91 | ||
More Information
Set Membership: |
|

Cdk14
Genomic Map