| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200011121.mm.1 |
NM_153198.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011121.mm.1 |
NM_153198.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011121.mm.1 |
TC1200001601.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200011124.mm.1 |
NM_153198.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011124.mm.1 |
NM_153198.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011124.mm.1 |
NM_177993.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011124.mm.1 |
NM_177993.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011124.mm.1 |
TC1200001601.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200011123.mm.1 |
NM_153198.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011123.mm.1 |
NM_153198.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011123.mm.1 |
NM_177993.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011123.mm.1 |
NM_177993.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200011123.mm.1 |
TC1200001601.mm.1 |
MTA TC |