Assay Details
| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146010.1 | NP_001139482.1 | 15 - 16 | 1778 | 112 | |
| NM_199012.2 | NP_950177.2 | 16 - 17 | 1850 | 112 | ||
| GenBank mRNA | AK122368.1 | - | 16 - 17 | 1742 | 112 | |
| AK140322.1 | - | 15 - 16 | 1757 | 112 | ||
| AK141949.1 | - | 15 - 16 | 1778 | 112 | ||
| BC039788.1 | - | 2 - 3 | 252 | 112 | ||
| BC060647.1 | - | 16 - 17 | 1812 | 6854041 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fchsd2
Genomic Map