Assay Details
| Gene Symbol: Psma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011965.2 | NP_036095.1 | 5 - 6 | 459 | 61 | |
| GenBank mRNA | AF060088.1 | - | 5 - 6 | 383 | 61 | |
| AJ272019.1 | - | 5 - 6 | 412 | 61 | ||
| AK088673.1 | - | 5 - 6 | 426 | 61 | ||
| AK137054.1 | - | 5 - 6 | 401 | 61 | ||
| AK144784.1 | - | 5 - 6 | 414 | 61 | ||
| AK161611.1 | - | 5 - 6 | 400 | 61 | ||
| AK162277.1 | - | 5 - 6 | 406 | 61 | ||
| BC005762.1 | - | 5 - 6 | 396 | 2655483 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psma1
Genomic Map