Assay Details
| Gene Symbol: Lsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110101.2 | NP_001103571.1 | 3 - 4 | 338 | 97 | |
| NM_001204274.1 | NP_001191203.1 | 3 - 4 | 338 | 97 | ||
| NM_030597.3 | NP_085100.1 | 3 - 4 | 344 | 97 | ||
| GenBank mRNA | AF204146.1 | - | 3 - 4 | 315 | 97 | |
| AF204147.1 | - | 3 - 4 | 315 | 97 | ||
| AF204148.1 | - | 3 - 4 | 315 | 97 | ||
| AF204149.1 | - | 3 - 4 | 315 | 97 | ||
| AF204150.1 | - | 3 - 4 | 315 | 97 | ||
| AF204151.1 | - | 3 - 4 | 315 | 97 | ||
| AF204152.1 | - | 3 - 4 | 315 | 97 | ||
| AF204153.1 | - | 2 - 3 | 557 | 97 | ||
| AF204157.1 | - | 3 - 4 | 182 | 97 | ||
| AK151605.1 | - | 3 - 4 | 312 | 97 | ||
| AK178855.1 | - | 3 - 4 | 126 | 97 | ||
| BB848823.1 | - | 3 - 4 | 344 | 97 | ||
| BC014288.1 | - | 3 - 4 | 323 | 3982410 | 97 | |
| BC049543.1 | - | 337 | 6774564 | 97 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lsm2
Genomic Map