Assay Details
Gene Symbol: Prl7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011168.4 | NP_035298.1 | 1 - 2 | 66 | 73 | |
GenBank mRNA | AF011382.1 | - | 1 - 2 | 55 | 73 | |
AF020524.1 | - | 1 - 2 | 62 | 73 | ||
AK005455.1 | - | 1 - 2 | 67 | 73 | ||
AK028068.1 | - | 1 - 2 | 66 | 72 | ||
AK131665.1 | - | 1 - 2 | 67 | 73 | ||
BC099468.1 | - | 1 - 2 | 45 | 30789902 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF011382.1, AF020524.1, AK005455.1, AK131665.1, BC099468.1, NM_011168.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
