Assay Details
| Gene Symbol: Lrrc51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027053.1 | NP_081329.1 | 2 - 3 | 156 | 104 | |
| GenBank mRNA | AK005758.1 | - | 2 - 3 | 107 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005758.1, NM_027053.1 transcripts.
Set Membership: |
|

Lrrc51
Genomic Map