Assay Details
| Gene Symbol: Lrrc51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162973.1 | NP_001156445.1 | 3 - 4 | 442 | 73 | |
| NM_001162974.1 | NP_001156446.1 | 2 - 3 | 348 | 73 | ||
| NM_027053.1 | NP_081329.1 | 4 - 5 | 505 | 73 | ||
| GenBank mRNA | AK005758.1 | - | 4 - 5 | 456 | 73 | |
| BC025128.1 | - | 2 - 3 | 399 | 4225298 | 73 | |
| EU627089.1 | - | 2 - 3 | 306 | 73 | ||
| EU627090.1 | - | 2 - 3 | 348 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999760 | NM_001162973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999760 | NM_001162974.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999760 | NM_027053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099627 | EU627089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099627 | EU627090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099627 | NM_001162973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099627 | NM_001162974.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099627 | NM_027053.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535596 | AK078038.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535596 | BC018249.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535596 | NM_011428.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168368 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87115 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80857 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112398 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130218 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099627 | EU627089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099627 | EU627090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099627 | NM_001162973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099627 | NM_001162974.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099627 | NM_027053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999760 | NM_001162973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999760 | NM_001162974.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999760 | NM_027053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319137 | EU627089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319137 | EU627090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319137 | NM_001162973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319137 | NM_001162974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P319137 | NM_027053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431510 | EU627090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431510 | NM_001162974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431510 | BC025128.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431510 | AK080275.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05933035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06653747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06147443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06532303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06532302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06027378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25376141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06091782 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566048 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566044 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566047 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566046 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566049 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429212_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427572_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566043 | - | - |
| Mouse Genome 430 2.0 Array | 1429212_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1429212_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69358_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69358_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700032477.mm.1 | NM_001162973.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032477.mm.1 | NM_001162973.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032477.mm.1 | NM_001162974.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032477.mm.1 | NM_001162974.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032477.mm.1 | NM_027053.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032477.mm.1 | NM_027053.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032477.mm.1 | TC0700003848.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032479.mm.1 | NM_001162973.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032479.mm.1 | NM_001162973.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032479.mm.1 | NM_001162974.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032479.mm.1 | NM_001162974.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032479.mm.1 | NM_027053.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032479.mm.1 | NM_027053.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032479.mm.1 | TC0700003848.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032481.mm.1 | NM_001162974.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032481.mm.1 | NM_001162974.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032481.mm.1 | TC0700003848.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032483.mm.1 | NM_001162973.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032483.mm.1 | NM_001162973.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032483.mm.1 | NM_001162974.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032483.mm.1 | NM_001162974.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032483.mm.1 | NM_027053.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032483.mm.1 | NM_027053.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032483.mm.1 | TC0700003848.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032482.mm.1 | NM_001162973.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032482.mm.1 | NM_001162973.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032482.mm.1 | NM_001162974.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032482.mm.1 | NM_001162974.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032482.mm.1 | NM_027053.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032482.mm.1 | NM_027053.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032482.mm.1 | TC0700003848.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427572_PM_at | - | - |
| Mouse Expression Set 430 | 1429212_a_at | - | - |
| Mouse Expression Set 430 | 1429212_PM_a_at | - | - |
| Mouse Expression Set 430 | 1427572_at | - | - |
| Murine Genome U74v2 Set | 135142_PM_at | - | - |
| Murine Genome U74v2 Set | 135142_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429212_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427572_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429212_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427572_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429212_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429212_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 580382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258770 | - | - |

Lrrc51
Genomic Map