| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00010852.mm.1 |
NM_001287530.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010852.mm.1 |
TC0X00001700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010853.mm.1 |
NM_001287530.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010853.mm.1 |
TC0X00001700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010851.mm.1 |
NM_178754.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010851.mm.1 |
NR_109847.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010851.mm.1 |
TC0X00001700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010855.mm.1 |
NM_001287530.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010855.mm.1 |
NM_009707.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010855.mm.1 |
NM_178754.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010855.mm.1 |
NR_109847.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010855.mm.1 |
TC0X00001700.mm.1 |
MTA TC |