Assay Details
| Gene Symbol: Arhgap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287530.1 | NP_001274459.1 | 3 - 4 | 581 | 83 | |
| NM_009707.4 | NP_033837.2 | 3 - 4 | 1949 | 83 | ||
| NM_178754.3 | NP_848869.1 | 3 - 4 | 492 | 83 | ||
| NR_109847.1 | 3 - 4 | 492 | 83 | |||
| GenBank mRNA | AK047827.1 | - | 3 - 4 | 493 | 83 | |
| AK048162.1 | - | 3 - 4 | 446 | 83 | ||
| AK048507.1 | - | 3 - 4 | 447 | 83 | ||
| AK077996.1 | - | 3 - 4 | 454 | 83 | ||
| AK079651.1 | - | 3 - 4 | 405 | 83 | ||
| AK164501.1 | - | 3 - 4 | 1950 | 83 | ||
| AK166015.1 | - | 3 - 4 | 412 | 83 | ||
| AK170575.1 | - | 3 - 4 | 447 | 83 | ||
| AK171337.1 | - | 3 - 4 | 378 | 83 | ||
| AK171677.1 | - | 3 - 4 | 1950 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap6
Genomic Map