Assay Details
| Gene Symbol: Klhl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161800.1 | NP_001155272.1 | 8 - 9 | 1342 | 94 | |
| NM_026448.3 | NP_080724.2 | 8 - 9 | 1342 | 94 | ||
| GenBank mRNA | AK012326.1 | - | 8 - 9 | 1323 | 94 | |
| AK083358.1 | - | 8 - 9 | 1328 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klhl7
Genomic Map