Assay Details
| Gene Symbol: Ly6g5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148939.2 | NP_683741.1 | 2 - 3 | 194 | 67 | |
| GenBank mRNA | AJ315553.1 | - | 2 - 3 | 194 | 67 | |
| AJ315554.1 | - | 1 - 2 | 290 | 67 | ||
| AK151902.1 | - | 1 - 2 | 231 | 67 | ||
| BC131964.1 | - | 2 - 3 | 222 | 40130241 | 67 | |
| BC131966.1 | - | 2 - 3 | 222 | 40130243 | 67 | |
| BC145150.1 | - | 1 - 2 | 318 | 67 | ||
| HE864438.1 | - | 2 - 3 | 210 | 67 | ||
| HE864439.1 | - | 1 - 2 | 306 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK151902.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972133 | AK151902.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972133 | BC131964.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972133 | BC131966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972133 | BC145150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972133 | NM_148939.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245765 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146921 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972133 | AK151902.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972133 | BC131964.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972133 | BC131966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972133 | BC145150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972133 | NM_148939.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369443 | AK151902.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369443 | BC131964.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369443 | BC131966.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369443 | BC145150.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369443 | NM_148939.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157656 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146921 | - | - |
| Whole Mouse Genome G4122A | A_51_P369443 | AK151902.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369443 | BC131964.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369443 | BC131966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369443 | BC145150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369443 | NM_148939.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111885 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146921 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505595 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505593 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505576 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13090435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13505595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13505582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13505600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13505573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157652 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450434 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450433 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450431 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700018473.mm.1 | NM_148939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018473.mm.1 | NM_148939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018473.mm.1 | TC1700001946.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018472.mm.1 | NM_148939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018472.mm.1 | NM_148939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018472.mm.1 | TC1700001946.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 352609 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6110129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608000 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2608000 | - | - |
| mouse-6 v1.1 (Illumina) | scl49997.2.1_244-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49997.2.1_244-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2608000 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2608000 | - | - |

Ly6g5b
Genomic Map