| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700036060.mm.1 |
NM_026827.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036060.mm.1 |
NM_026827.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036060.mm.1 |
NM_028389.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036060.mm.1 |
NM_028389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036060.mm.1 |
TC0700004303.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036063.mm.1 |
TC0700004303.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036059.mm.1 |
NM_026827.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036059.mm.1 |
NM_026827.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036059.mm.1 |
NM_028389.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036059.mm.1 |
NM_028389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036059.mm.1 |
TC0700004303.mm.1 |
MTA TC |