Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026827.1 | NP_081103.1 | 3 - 4 | 445 | 75 | |
| NM_028389.1 | NP_082665.1 | 3 - 4 | 442 | 75 | ||
| GenBank mRNA | AK004043.1 | - | 2 - 3 | 405 | 75 | |
| AK013646.1 | - | 2 - 3 | 247 | 75 | ||
| AK152526.1 | - | 3 - 4 | 407 | 75 | ||
| BC023442.1 | - | 3 - 4 | 420 | 5026775 | 75 | |
| BC046763.1 | - | 2 - 3 | 587 | 5693779 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem219
Genomic Map