Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026827.1 | NP_081103.1 | 4 - 5 | 677 | 78 | |
| NM_028389.1 | NP_082665.1 | 4 - 5 | 674 | 78 | ||
| GenBank mRNA | AK004043.1 | - | 3 - 4 | 637 | 78 | |
| AK152526.1 | - | 4 - 5 | 639 | 78 | ||
| BC023442.1 | - | 4 - 5 | 652 | 5026775 | 78 | |
| BC046763.1 | - | 3 - 4 | 819 | 5693779 | 78 | |
| BP769695.1 | - | 2 - 3 | 78 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091486 | BC046763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091486 | BP769695.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091486 | BC023442.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091486 | NM_026827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091486 | NM_028389.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421385 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78265 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110872 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153537 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091486 | BC046763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091486 | BP769695.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091486 | BC023442.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091486 | NM_026827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091486 | NM_028389.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110872 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06360216 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153537 | - | - |
| Whole Mouse Genome G4122A | A_52_P225466 | BC046763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P225466 | BC023442.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P225466 | NM_026827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P225466 | NM_028389.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P225466 | AK004043.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454958 | BC046763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454958 | BP769695.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454958 | BC023442.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454958 | NM_026827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P454958 | NM_028389.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153537 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110872 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06401336 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06401331 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06401324 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06401327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05975173 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06401326 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06401335 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06401332 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06603048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25502812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06444744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06401326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06558305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06401332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06444003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06089533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06444745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06401335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06069844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06401331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06305936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06211878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05954886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05967437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06318152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06630479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05975173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05893984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06294906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06368531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06444001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05992990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06305934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05893985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06486639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06443998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25502795 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568106 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430505_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457243_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424497_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568099 | - | - |
| Mouse Genome 430 2.0 Array | 1424497_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457243_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424497_at | - | - |
| Mouse Genome 430 2.0 Array | 1430505_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430505_at | - | - |
| Mouse Genome 430 2.0 Array | 1457243_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68742_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68742_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700036054.mm.1 | NM_026827.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036054.mm.1 | NM_026827.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036054.mm.1 | NM_028389.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036054.mm.1 | NM_028389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036054.mm.1 | TC0700004303.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036052.mm.1 | NM_026827.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036052.mm.1 | NM_026827.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036052.mm.1 | NM_028389.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036052.mm.1 | NM_028389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036052.mm.1 | TC0700004303.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036053.mm.1 | NM_026827.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036053.mm.1 | NM_026827.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036053.mm.1 | NM_028389.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036053.mm.1 | NM_028389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036053.mm.1 | TC0700004303.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424497_PM_at | - | - |
| Mouse Expression Set 430 | 1424497_at | - | - |
| Mouse Expression Set 430 | 1457243_at | - | - |
| Mouse Expression Set 430 | 1457243_PM_at | - | - |
| Murine Genome U74v2 Set | 169719_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169719_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424497_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424497_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424497_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424497_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457243_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430505_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457243_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430505_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 904864 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689569 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689569 | - | - |

Tmem219
Genomic Map