Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026827.1 | NP_081103.1 | 5 - 6 | 64 | ||
| NM_028389.1 | NP_082665.1 | 5 - 6 | 64 | |||
| GenBank mRNA | AK004043.1 | - | 4 - 5 | 64 | ||
| AK152526.1 | - | 5 - 6 | 64 | |||
| BC023442.1 | - | 5 - 6 | 5026775 | 64 | ||
| BC046763.1 | - | 4 - 5 | 5693779 | 64 | ||
| BP769695.1 | - | 1 - 2 | 119 | 64 | ||
More Information
Set Membership: |
|

Tmem219
Genomic Map