Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026827.1 | NP_081103.1 | 1 - 2 | 57 | 106 | |
| GenBank mRNA | BC023442.1 | - | 1 - 2 | 32 | 5026775 | 106 |
| BY128578.1 | - | 1 - 2 | 57 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023442.1, NM_026827.1 transcripts.
Set Membership: |
|

Tmem219
Genomic Map