Assay Details
| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030256.2 | NP_084532.2 | 2 - 3 | 241 | 66 | |
| GenBank mRNA | AK155681.1 | - | 3 - 4 | 201 | 66 | |
| AY296058.1 | - | 2 - 3 | 241 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY296058.1, NM_030256.2 transcripts.
Set Membership: |
|

Bcl9l
Genomic Map