| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000006059.mm.1 |
NM_001288625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006059.mm.1 |
NM_001288626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006059.mm.1 |
NM_007880.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006059.mm.1 |
TC1000000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000006055.mm.1 |
NM_001288626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006055.mm.1 |
NM_007880.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006055.mm.1 |
TC1000000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000006060.mm.1 |
NM_001288625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006060.mm.1 |
NM_001288626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006060.mm.1 |
NM_007880.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006060.mm.1 |
TC1000000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000006053.mm.1 |
NM_001288626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006053.mm.1 |
NM_007880.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006053.mm.1 |
TC1000000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000006054.mm.1 |
NM_001288626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006054.mm.1 |
NM_007880.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006054.mm.1 |
TC1000000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000006056.mm.1 |
NM_001288625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006056.mm.1 |
TC1000000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000006058.mm.1 |
NM_001288625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006058.mm.1 |
NM_001288626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006058.mm.1 |
NM_007880.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000006058.mm.1 |
TC1000000829.mm.1 |
MTA TC |