Assay Details
| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172926.3 | NP_766514.2 | 23 - 24 | 2505 | 112 | |
| GenBank mRNA | AK049371.1 | - | 23 - 24 | 2484 | 112 | |
| AK082389.1 | - | 23 - 24 | 2426 | 112 | ||
| BC043328.1 | - | 8 - 9 | 1163 | 112 | ||
| BC080799.1 | - | 23 - 24 | 2490 | 4948538 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx14
Genomic Map