Assay Details
| Gene Symbol: Pgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163746.1 | NP_001157218.1 | 2 - 3 | 319 | 65 | |
| NM_028352.4 | NP_082628.3 | 2 - 3 | 319 | 65 | ||
| GenBank mRNA | AK013402.1 | - | 2 - 3 | 302 | 65 | |
| AK028066.1 | - | 2 - 3 | 309 | 65 | ||
| AK032776.1 | - | 2 - 3 | 271 | 65 | ||
| AK036561.1 | - | 2 - 3 | 305 | 65 | ||
| AK043547.1 | - | 2 - 3 | 320 | 65 | ||
| AK049337.1 | - | 2 - 3 | 271 | 65 | ||
| AK049758.1 | - | 2 - 3 | 306 | 65 | ||
| AK051706.1 | - | 2 - 3 | 259 | 65 | ||
| AK153501.1 | - | 1 - 2 | 139 | 65 | ||
| AK160913.1 | - | 2 - 3 | 320 | 65 | ||
| AK165513.1 | - | 2 - 3 | 304 | 65 | ||
| AK169082.1 | - | 2 - 3 | 284 | 65 | ||
| AK169141.1 | - | 2 - 3 | 278 | 65 | ||
| AK169787.1 | - | 2 - 3 | 293 | 65 | ||
| BC138700.1 | - | 2 - 3 | 244 | 8861722 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgm3
Genomic Map