Assay Details
| Gene Symbol: Lppr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170935.1 | NP_001164406.1 | 6 - 7 | 1369 | 83 | |
| NM_181681.2 | NP_859009.2 | 6 - 7 | 1369 | 83 | ||
| NR_033210.1 | 5 - 6 | 1510 | 83 | |||
| GenBank mRNA | AF541278.1 | - | 6 - 7 | 1369 | 83 | |
| AK083932.1 | - | 5 - 6 | 1167 | 83 | ||
| AK144442.1 | - | 6 - 7 | 1047 | 83 | ||
| AK220474.1 | - | 5 - 6 | 1446 | 83 | ||
| BC005764.1 | - | 6 - 7 | 1124 | 83 | ||
| BC057042.1 | - | 1 - 2 | 163 | 83 | ||
| BC066006.1 | - | 6 - 7 | 964 | 6808849 | 83 | |
| BC098099.1 | - | 6 - 7 | 1122 | 6813259 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279424 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279423 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060991 | BC079534.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060991 | BC005764.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060991 | BC057042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060991 | BC066006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060991 | AK144442.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117721 | BC098099.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117721 | NM_001170935.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719484 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61530 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80868 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107699 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117721 | BC098099.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117721 | NM_001170935.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060991 | BC079534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060991 | BC005764.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060991 | BC057042.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060991 | BC066006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060991 | AK144442.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279422 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107699 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02167885 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07120680 | - | - |
| Whole Mouse Genome G4122A | A_51_P186178 | BC079534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P186178 | BC005764.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P186178 | BC057042.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P186178 | BC066006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P186178 | AK144442.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123438 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107699 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08818169 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08372334 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08372325 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08496355 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08818168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08496357 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08372318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08372327 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279424 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08452789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08886265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08610980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08354783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08337261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08818169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08634911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08886267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08700409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08740111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279437 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10370661 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370655 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370657 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370660 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370662 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370658 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370664 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370663 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454761_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10370651 | - | - |
| Mouse Genome 430 2.0 Array | 1454761_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454761_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216152_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216152_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000018164.mm.1 | NM_001170935.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018164.mm.1 | NM_001170935.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018164.mm.1 | NM_181681.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018164.mm.1 | NM_181681.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018164.mm.1 | NR_033210.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018164.mm.1 | NR_033210.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018164.mm.1 | TC1000002482.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018163.mm.1 | NM_181681.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018163.mm.1 | NM_181681.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018163.mm.1 | NR_033210.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018163.mm.1 | NR_033210.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018163.mm.1 | TC1000002482.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018161.mm.1 | NM_001170935.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018161.mm.1 | NM_001170935.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018161.mm.1 | TC1000002482.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018162.mm.1 | NM_001170935.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018162.mm.1 | NM_001170935.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018162.mm.1 | TC1000002482.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454761_PM_at | - | - |
| Mouse Expression Set 430 | 1454761_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454761_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454761_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 106660270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6020152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2709001 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2709001 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2576574 | - | - |
| mouse-6 v1.1 (Illumina) | scl37747.5_571-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106660270 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020152 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37747.5_571-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106660270 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2709001 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252887 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2709001 | - | - |

Lppr3
Genomic Map