Assay Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144528.3 | NP_653111.1 | 5 - 6 | 636 | 90 | |
| GenBank mRNA | AK154781.1 | - | 5 - 6 | 602 | 90 | |
| BC016543.1 | - | 5 - 6 | 608 | 3158083 | 90 | |
| CA321804.1 | - | 2 - 3 | 143 | 90 | ||
| DV648665.1 | - | 5 - 6 | 531 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf126
Genomic Map