| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600004884.mm.1 |
NM_001285788.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004884.mm.1 |
NM_001285791.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004884.mm.1 |
NM_134109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004884.mm.1 |
TC1600000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004883.mm.1 |
NM_001285788.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004883.mm.1 |
NM_001285791.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004883.mm.1 |
NM_134109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004883.mm.1 |
TC1600000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004881.mm.1 |
NM_001285788.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004881.mm.1 |
NM_001285791.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004881.mm.1 |
NM_134109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004881.mm.1 |
TC1600000522.mm.1 |
MTA TC |