Assay Details
| Gene Symbol: Lca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027448.2 | NP_081724.1 | 3 - 4 | 1089 | 93 | |
| NM_029434.3 | NP_083710.2 | 3 - 4 | 985 | 93 | ||
| GenBank mRNA | AK015260.1 | - | 3 - 4 | 957 | 93 | |
| BC049101.1 | - | 3 - 4 | 1017 | 5702365 | 93 | |
| BC052060.1 | - | 3 - 4 | 1017 | 5702365 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lca5
Genomic Map