Assay Details
| Gene Symbol: Bace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145947.1 | NP_001139419.1 | 2 - 3 | 852 | 62 | |
| NM_011792.5 | NP_035922.4 | 2 - 3 | 852 | 62 | ||
| GenBank mRNA | AF190726.2 | - | 2 - 3 | 780 | 62 | |
| AF200346.1 | - | 2 - 3 | 353 | 62 | ||
| AK014464.1 | - | 2 - 3 | 782 | 62 | ||
| AK033112.1 | - | 2 - 3 | 802 | 62 | ||
| AK041285.1 | - | 2 - 3 | 802 | 62 | ||
| AK046175.1 | - | 2 - 3 | 799 | 62 | ||
| AK049626.1 | - | 2 - 3 | 785 | 62 | ||
| AK080498.1 | - | 2 - 3 | 804 | 62 | ||
| AK082230.1 | - | 2 - 3 | 800 | 62 | ||
| AK082317.1 | - | 2 - 3 | 803 | 62 | ||
| AK163583.1 | - | 2 - 3 | 803 | 62 | ||
| BC048189.1 | - | 2 - 3 | 778 | 6831622 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bace1
Genomic Map