| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700024911.mm.1 |
NM_001164184.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024911.mm.1 |
NM_001164184.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024911.mm.1 |
NM_001164185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024911.mm.1 |
NM_001164185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024911.mm.1 |
NM_017405.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024911.mm.1 |
NM_017405.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024911.mm.1 |
TC0700002785.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700024912.mm.1 |
NM_001164184.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024912.mm.1 |
NM_001164184.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024912.mm.1 |
NM_001164185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024912.mm.1 |
NM_001164185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024912.mm.1 |
NM_017405.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024912.mm.1 |
NM_017405.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024912.mm.1 |
TC0700002785.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700024910.mm.1 |
NM_001164184.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024910.mm.1 |
NM_001164184.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024910.mm.1 |
NM_001164185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024910.mm.1 |
NM_001164185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024910.mm.1 |
NM_017405.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024910.mm.1 |
NM_017405.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024910.mm.1 |
TC0700002785.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700024915.mm.1 |
NM_001164184.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024915.mm.1 |
NM_001164184.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024915.mm.1 |
NM_001164185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024915.mm.1 |
NM_001164185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024915.mm.1 |
NM_017405.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024915.mm.1 |
NM_017405.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700024915.mm.1 |
TC0700002785.mm.1 |
MTA TC |