Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900000746.mm.1 |
NM_023153.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000746.mm.1 |
NM_023153.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000746.mm.1 |
TC0900000105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000744.mm.1 |
NM_023153.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000744.mm.1 |
NM_023153.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000744.mm.1 |
TC0900000105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000747.mm.1 |
NM_023153.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000747.mm.1 |
NM_023153.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000747.mm.1 |
TC0900000105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000742.mm.1 |
NM_023153.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000742.mm.1 |
NM_023153.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000742.mm.1 |
TC0900000105.mm.1 |
MTA TC |