Assay Details
| Gene Symbol: Slc19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199271.1 | NP_001186200.1 | 2 - 3 | 270 | 81 | |
| NM_031196.3 | NP_112473.1 | 2 - 3 | 364 | 81 | ||
| GenBank mRNA | AK002560.1 | - | 2 - 3 | 281 | 81 | |
| AK050714.1 | - | 2 - 3 | 394 | 81 | ||
| AK079592.1 | - | 2 - 3 | 325 | 81 | ||
| AK085024.2 | - | 2 - 3 | 310 | 81 | ||
| AK088803.1 | - | 2 - 3 | 327 | 81 | ||
| AK138577.1 | - | 2 - 3 | 325 | 81 | ||
| BC015263.1 | - | 2 - 3 | 282 | 4239690 | 81 | |
| BF786711.1 | - | 2 - 3 | 281 | 81 | ||
| L23755.1 | - | 1 - 2 | 222 | 81 | ||
| L36539.1 | - | 2 - 3 | 277 | 81 | ||
| U32469.1 | - | 2 - 3 | 304 | 81 | ||
| U66103.1 | - | 2 - 3 | 317 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc19a1
Genomic Map