| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700022675.mm.1 |
NM_207677.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022675.mm.1 |
NM_207677.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022675.mm.1 |
TC0700002625.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700022679.mm.1 |
NM_207677.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022679.mm.1 |
NM_207677.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022679.mm.1 |
TC0700002625.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700022678.mm.1 |
NM_207677.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022678.mm.1 |
NM_207677.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022678.mm.1 |
TC0700002625.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700022682.mm.1 |
NM_207677.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022682.mm.1 |
NM_207677.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022682.mm.1 |
TC0700002625.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700022677.mm.1 |
NM_207677.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022677.mm.1 |
NM_207677.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700022677.mm.1 |
TC0700002625.mm.1 |
MTA TC |