Assay Details
| Gene Symbol: Ggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079235.1 | - | 7 - 8 | 983 | 76 | |
| BC012969.1 | - | 6 - 7 | 872 | 4037243 | 76 | |
| CT010196.1 | - | 5 - 6 | 730 | 76 | ||
| U30509.1 | - | 6 - 7 | 890 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ggt1
Genomic Map