Assay Details
| Gene Symbol: Ggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC012969.1 | - | 8 - 9 | 1166 | 4037243 | 66 |
| CT010196.1 | - | 7 - 8 | 1024 | 66 | ||
| U30509.1 | - | 8 - 9 | 1184 | 66 | ||
More Information
Set Membership: |
|

Ggt1
Genomic Map